Tisk…

Anti-Growth Hormone Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB24389-1EA 0 CZK
BIRBORB24389-1
Anti-Growth Hormone Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Growth Hormone
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR