Tisk…

Anti-DNP Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB24458-100EA 0 CZK
BIRBORB24458-100
Anti-DNP Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: DNP (Dinitrophenol)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR