Tisk…

Anti-PEG Mouse Monoclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB24468-50EA 0 CZK
BIRBORB24468-50
Anti-PEG Mouse Monoclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: PEG
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR