Tisk…
Anti-Amitriptyline Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB24547-0.2EA 0 CZK
BIRBORB24547-0.2
Anti-Amitriptyline Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Amitriptyline
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat