Tisk…

Anti-B2M Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB24720-0.2EA 0 CZK
BIRBORB24720-0.2
Anti-B2M Mouse Monoclonal Antibody
Protilátky
  • Flow Cytometry: 2 μg; Immunochemistry: 1:25-1:50

Type: Primary
Antigen: B2M (beta-2-microglobulin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR