Tisk…
Anti-Alpha 1-Antitrypsin Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB24734-1EA 0 CZK
BIRBORB24734-1
Anti-Alpha 1-Antitrypsin Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Alpha 1-Antitrypsin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Objednat