Tisk…

Anti-Chymotrypsin Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB24774-0.2EA 0 CZK
BIRBORB24774-0.2
Anti-Chymotrypsin Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: Chymotrypsin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR