Tisk…

Anti-proBNP Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB24884-1EA 0 CZK
BIRBORB24884-1
Anti-proBNP Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: proBNP
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR