Tisk…

Anti-Ferritin Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB24932-1EA 0 CZK
BIRBORB24932-1
Anti-Ferritin Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Ferritin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR