Tisk…

Anti-EGFR (External Domain) Mouse Monoclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB25007-0.25EA 0 CZK
BIRBORB25007-0.25
Anti-EGFR (External Domain) Mouse Monoclonal Antibody (Biotin)
Protilátky
Flow Cytometry: 1 ?g-1.0 x106

Type: Primary
Antigen: EGFR (External Domain)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR