Tisk…

Anti-Sm Antigen Human Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25156-2EA 0 CZK
BIRBORB25156-2
Anti-Sm Antigen Human Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Sm Antigen
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Human
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR