Tisk…

Anti-Hamster Apo E Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25176-1EA 0 CZK
BIRBORB25176-1
Anti-Hamster Apo E Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Hamster Apo E
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Hamster
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR