Tisk…

Anti-NPR2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25187-20EA 0 CZK
BIRBORB25187-20
Anti-NPR2 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:10000

Type: Primary
Antigen: NPR2 (natriuretic peptide receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR