Tisk…

Anti-HOCl-OX LDL Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25189-100EA 0 CZK
BIRBORB25189-100
Anti-HOCl-OX LDL Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:400-1:4000; Immunochemistry: 1:400

Type: Primary
Antigen: HOCl-OX LDL
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR