Tisk…

Anti-IL17A Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25229-100EA 0 CZK
BIRBORB25229-100
Anti-IL17A Rabbit Polyclonal Antibody
Protilátky
ELISA: 100 ?l; Western Blot: 1.5-3.0 ng

Type: Primary
Antigen: IL17A (interleukin 17A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR