Tisk…

Anti-APOB Goat Polyclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB25255-1EA 0 CZK
BIRBORB25255-1
Anti-APOB Goat Polyclonal Antibody (Biotin)
Protilátky
ELISA: 1:200-1:40000

Type: Primary
Antigen: APOB (apolipoprotein B)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR