Tisk…
Anti-Alk. Phos. (APAAP) Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB25272-1EA 0 CZK
BIRBORB25272-1
Anti-Alk. Phos. (APAAP) Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Alk. Phos. (APAAP)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat