Tisk…

Anti-NPPA Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25298-100EA 0 CZK
BIRBORB25298-100
Anti-NPPA Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NPPA (natriuretic peptide A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR