Tisk…

Anti-PSAT1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25308-1EA 0 CZK
BIRBORB25308-1
Anti-PSAT1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PSAT1 (phosphoserine aminotransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR