Tisk…

Anti-CUL2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25426-100EA 0 CZK
BIRBORB25426-100
Anti-CUL2 Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:2000-1:10000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: CUL2 (cullin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR