Tisk…

Anti-ALB Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25467-10EA 0 CZK
BIRBORB25467-10
Anti-ALB Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ALB (albumin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR