Tisk…

Anti-PAI-1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25479-0.2EA 0 CZK
BIRBORB25479-0.2
Anti-PAI-1 Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:50000; Western Blot: 1:5000; Immunochemistry: 1:500

Type: Primary
Antigen: PAI-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR