Tisk…

Anti-PLAU Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25484-0.2EA 0 CZK
BIRBORB25484-0.2
Anti-PLAU Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:10000; Western Blot: 1:5000

Type: Primary
Antigen: PLAU (plasminogen activator, urokinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR