Tisk…

Anti-EGFR Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB25496-100EA 0 CZK
BIRBORB25496-100
Anti-EGFR Mouse monoclonal antibody
Protilátky
DB: 0.5 ?g/ml; ELISA: 0.05 ?g/ml

Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR