Tisk…

Anti-APOC1 Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25546-1EA 0 CZK
BIRBORB25546-1
Anti-APOC1 Goat Polyclonal Antibody
Protilátky
ELISA: 1:20000-1:80000

Type: Primary
Antigen: APOC1 (apolipoprotein C-I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR