Tisk…

Anti-APOC3 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Biorbyt
BIRBORB25552-1EA 0 CZK
BIRBORB25552-1
Anti-APOC3 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
ELISA: 1:500-1:6000

Type: Primary
Antigen: APOC3 (apolipoprotein C-III)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR