Tisk…

Anti-SERPINC1 Sheep Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25717-1EA 0 CZK
BIRBORB25717-1
Anti-SERPINC1 Sheep Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SERPINC1 (serpin peptidase inhibitor, clade C (antithrombin), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR