Tisk…

Anti-APOC2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25778-1EA 0 CZK
BIRBORB25778-1
Anti-APOC2 Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:2000-1:8000

Type: Primary
Antigen: APOC2 (apolipoprotein C-II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR