Tisk…

Anti-APOA2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB25785-0.5EA 0 CZK
BIRBORB25785-0.5
Anti-APOA2 Rabbit Polyclonal Antibody
Protilátky
ELISAl: 1:200-1:20000

Type: Primary
Antigen: APOA2 (apolipoprotein A-II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR