Tisk…

Anti-MUC1 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB25897-1EA 0 CZK
BIRBORB25897-1
Anti-MUC1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: MUC1 (mucin 1, cell surface associated)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR