Tisk…

Anti-PSPN Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB25913-0.5EA 0 CZK
BIRBORB25913-0.5
Anti-PSPN Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PSPN (persephin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR