Tisk…

Anti-COL3 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB25918-100EA 0 CZK
BIRBORB25918-100
Anti-COL3 Mouse Monoclonal Antibody
Protilátky
ELISA: 1:64000; Immunochemistry: 1:200

Type: Primary
Antigen: COL3 (collagen, type III)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR