Tisk…

Anti-COL1 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB25932-100EA 0 CZK
BIRBORB25932-100
Anti-COL1 Mouse Monoclonal Antibody
Protilátky
Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR