Tisk…

Anti-EPCAM Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB25961-50EA 0 CZK
BIRBORB25961-50
Anti-EPCAM Mouse Monoclonal Antibody
Protilátky
Immunochemistry: 1:10

Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR