Tisk…
Anti-Neuron Specific Enolase Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB26001-1EA 0 CZK
BIRBORB26001-1
Anti-Neuron Specific Enolase Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Neuron Specific Enolase
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human