Tisk…
Anti-CA125, Group B Mouse monoclonal antibody
Dodavatel: Biorbyt
BIRBORB26035-1EA 0 CZK
BIRBORB26035-1
Anti-CA125, Group B Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: CA125, Group B
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat