Tisk…

Anti-hGH Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26077-1EA 0 CZK
BIRBORB26077-1
Anti-hGH Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: hGH
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR