Tisk…

Anti-NCAM1 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26121-100EA 0 CZK
BIRBORB26121-100
Anti-NCAM1 Mouse Monoclonal Antibody
Protilátky
Immunochemistry: 1:50

Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR