Tisk…

Anti-AFP Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26138-1EA 0 CZK
BIRBORB26138-1
Anti-AFP Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: AFP (alpha-fetoprotein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR