Tisk…

Anti-FTH1 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26147-1EA 0 CZK
BIRBORB26147-1
Anti-FTH1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: FTH1 (ferritin, heavy polypeptide 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR