Tisk…

Anti-DES Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB26154-100EA 0 CZK
BIRBORB26154-100
Anti-DES Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: DES (desmin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR