Tisk…

Anti-FUT4 Mouse Monoclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB26226-100EA 0 CZK
BIRBORB26226-100
Anti-FUT4 Mouse Monoclonal Antibody (Biotin)
Protilátky
Flow Cytometry: 1:1000

Type: Primary
Antigen: FUT4 (fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific))
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR