Tisk…

Anti-Mouse Factor X Rat Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26258-100EA 0 CZK
BIRBORB26258-100
Anti-Mouse Factor X Rat Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Mouse Factor X
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR