Tisk…

Anti-D-Dimer Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26266-1EA 0 CZK
BIRBORB26266-1
Anti-D-Dimer Monoclonal Antibody
Protilátky
Type: Primary
Antigen: D-Dimer
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR