Tisk…

Anti-F8 Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB26278-0.2EA 0 CZK
BIRBORB26278-0.2
Anti-F8 Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: F8 (coagulation factor VIII, procoagulant component)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR