Tisk…

Anti-CD15 Granulocytes Mouse Monoclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB26358-0.2EA 0 CZK
BIRBORB26358-0.2
Anti-CD15 Granulocytes Mouse Monoclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: CD15 Granulocytes
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR