Tisk…

Anti-CCR5 Rat Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26394-0.2EA 0 CZK
BIRBORB26394-0.2
Anti-CCR5 Rat Monoclonal Antibody
Protilátky
Flow Cytometry: 1:50-1:100

Type: Primary
Antigen: CCR5 (chemokine (C-C motif) receptor 5 (gene/pseudogene))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR