Tisk…

Anti-Mouse CD54 (ICAM-1) Rat Monoclonal Antibody (PE (Phycoerythrin))

Dodavatel: Biorbyt
BIRBORB26419-100EA 0 CZK
BIRBORB26419-100
Anti-Mouse CD54 (ICAM-1) Rat Monoclonal Antibody (PE (Phycoerythrin))
Protilátky
Type: Primary
Antigen: Mouse CD54 (ICAM-1)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a (Rat)
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR