Tisk…

Anti-DEFB1 Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB26479-10EA 0 CZK
BIRBORB26479-10
Anti-DEFB1 Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: DEFB1 (defensin, beta 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR