Tisk…

Anti-IL-15 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB26484-0.5EA 0 CZK
BIRBORB26484-0.5
Anti-IL-15 Mouse Monoclonal Antibody
Protilátky
ELISA: 1:20000

Type: Primary
Antigen: IL-15
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR